
4-[3-(Trifluoromethyl)phenyl]-4-piperidinol 97%
Catalog # ACRO136980250
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Light beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:91 to 95 °C
- GC:≥96.0 %
- Elemental Analysis:5.73 to 6.21 % N
Specification Test Results
About this item
:
Melting Pt:91…95 °C
:245.245 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:2249-28-7
Formula:C₁₂H₁₄F₃NO
MDL Number:MFCD00044832