Order Entry
Netherlands
ContactUsLinkComponent
Rutin hydrate 97+%
Rutin hydrate 97+%
Certificates
undefined
Rutin hydrate 97+%
Certificates
Specification Test Results
  • Appearance:
    Yellow to yellow-green to straw yellow Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    5.0 % to 9.0 % (500 mg, 105°C, 3 h)
  • Ash:
    ≤0.5 %
  • Titration with NaOH:
    97.0 to 102.0 % (On dry basis)