1,2:5,6-Di-O-isopropylidene-α-D-glucofuranose 98+%
Leverancier: Thermo Fisher Scientific
Certificaten
Over dit artikel
CAS-nummer:582-52-5
:
Formule:C₁₂H₂₀O₆
MDL-nummer:MFCD00005544
:260.29 g/mol
Smelten Pt:107…110 °C
Temperatuur bij opslag:Freezer
Specificatie testresultaten
- Appearance:White to yellow Powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 %
- Sulfated ash:≤0.2 %
- GC:≥98.0 %
- Specific optical rotation:-10° to -12° (20°C, 589 nm) (c=5, EtOH)
Specificatie testresultaten
Frequently Bought Together