

Methyl-4-(chloroformyl)benzoate 90% tech.
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7377-26-6
:
Formula:C₉H₇ClO₃
MDL Number:MFCD00059486
:198.60 g/mol
Melting Pt:50…59 °C
UN:3261
ADR:8,II
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Off-white to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:50 to 59 °C
- Titration RCOCl:≥88 %
Specification Test Results
Frequently Bought Together