Sodium palmitate 98%
:408-35-5
:
:C₁₆H₃₂O₂
:MFCD00002749
:278.41 g/mol
:Ambient
- Appearance:White Powder
- Infrared spectrum:Conforms
- Loss on drying:≤2 % (110°C, 4 hrs)
- Titration with HClO4:97.5 to 102.5 % (On dry substance)
Frequently Bought Together