Order Entry
Netherlands
ContactUsLinkComponent
Hexadecyltrimethylammonium hydroxide 25% in methanol
Toxic
Health
Corrosive
Flammable
Hexadecyltrimethylammonium hydroxide 25% in methanol
Certificates
undefined
Toxic
Health
Corrosive
Flammable
Hexadecyltrimethylammonium hydroxide 25% in methanol

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Solution
  • Water:
    ≤3 % (K.F.)
  • Concentration:
    23 to 27 w/w% (in methanol)
  • Enantiomeric excess:
    ≥97.5 % (GC)

Specification Test Results

Related Information

Frequently Bought Together