
Chlorazol black E, pure, certified
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1937-37-7
:
Formula:C₃₄H₂₇N₉O₇S₂
MDL Number:MFCD00066363
:781.73 g/mol
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Dark brown to black Crystalline powder
- Infrared spectrum:Conforms
- UV Lambda max:597 to 602 nm
- Absorption Ratio P-15/P+15:0.97 to 1.00
- BSC Certification:Passes
Specification Test Results
Frequently Bought Together