Order Entry
Netherlands
ContactUsLinkComponent
Bis(2,4,6-trichlorophenyl)oxalate 98%
Irritant
Bis(2,4,6-trichlorophenyl)oxalate 98%
Catalog # ACRO402660050
CAS Number:  
undefined
Irritant
Bis(2,4,6-trichlorophenyl)oxalate 98%
Catalog # ACRO402660050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White Powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    187 to 193 °C
  • Titration with TBAH:
    ≥97.5 %

Specification Test Results

About this item

 :
 :448.88 g/mol
Melting Pt:188…192 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:1165-91-9
Formula:C₁₄H₄Cl₆O₄
MDL Number:MFCD00043061