


Benzyldimethylhexadecylammonium chloride hydrate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:122-18-9
:
Formula:C₂₅H₄₆ClN
MDL Number:MFCD00149967
Melting Pt:54…58 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS05
:GHS07
:GHS09
Specification Test Results
- Appearance:White to cream Crystalline powder
- Infrared spectrum:Conforms
- Titration Argentometric:≥96.0 % (on anhydrous substance)
- Water:≤10 % (K.F.)
Specification Test Results
Frequently Bought Together