Piperonyl butoxide 90% tech.
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:51-03-6
:
Formula:C₁₉H₃₀O₅
MDL Number:MFCD00005842
:338.44 g/mol
Boiling Pt:180 °C (1 mmHg)
Flash Pt:170 °C
Storage Temperature:Ambient
UN:3082
ADR:9,III
:GHS09
Specification Test Results
- Appearance:Clear yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4970 to 1.5070 (20°C, 589 nm)
- GC:≥88 %
- Specific gravity:(20°C) 1.055 to 1.065
Specification Test Results
Frequently Bought Together