Order Entry
Netherlands
ContactUsLinkComponent
2-Nitrophenyl selenocyanate 98%
Toxic
Health
Environment
2-Nitrophenyl selenocyanate 98%
Catalog # ACRO293580010
CAS Number:  
undefined
Toxic
Health
Environment
2-Nitrophenyl selenocyanate 98%
Catalog # ACRO293580010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Yellow to brown Powder or crystals or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    139 to 145 °C
  • GC:
    ≥97.5 %

Specification Test Results

About this item

 :
 :227.092 g/mol
Melting Pt:138…142 °C
Storage Temperature:Ambient
UN:3283
ADR:6.1,III
 :GHS06
 :GHS08
 :GHS09
CAS Number:51694-22-5
Formula:C₇H₄N₂O₂Se
MDL Number:MFCD00043146