
(-)-Terpinen-4-ol 97% (sum of enantiomers)
Leverancier: Thermo Fisher Scientific
Certificaten
Over dit artikel
CAS-nummer:20126-76-5
:
Formule:C₁₀H₁₈O
MDL-nummer:MFCD00167108
:154.25 g/mol
Kokend Pt:212 °C (760 mmHg)
Dichtheid:0.933 g/cm³
Flits Pt:90 °C
:GHS07
Temperatuur bij opslag:Ambient
Specificatie testresultaten
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4760 to 1.4820 (20°C, 589 nm)
- Density:(20/4°C) 0.930 to 0.936 g/ml
- GC:≥96.0 % (sum of enantiomers)
- Specific optical rotation:-19° to -31° (20°C, 589 nm) (neat)
Specificatie testresultaten
Frequently Bought Together