L-Valine methyl ester hydrochloride 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6306-52-1
:
Formula:C₆H₁₄ClNO₂
MDL Number:MFCD00012497
:167.63 g/mol
Melting Pt:171…173 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Powder or crystals
- Infrared spectrum:Conforms
- Titration Argentometric:≥98.5 %
- Solubility:(5 % in water) Clear colorless to light yellow
- Specific optical rotation:+14° to +17° (20°C, 589 nm) (c=2, water)
Specification Test Results
Frequently Bought Together