Anti-IgG Goat Polyclonal Antibody (APC (Allophycocyanin))
Supplier: Biotium
Certificates
About this item
This product is prepared by labeling goat anti-rat IgG (H+L) with allophycocyanin (APC). APC belongs to the family of phycobiliproteins, which are derived from cyanobacteria and eukaryotic algae. APC has absorption maxima at 650 and emission at 660 nm. Because of the high fluorescence quantum yield (~0.68), APC is by far the brightest dye for the 650 nm wavelength region.To minimize crossreactivity, the antibody has been adsorbed against human, bovine, horse, and rabbit serum proteins.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: APC
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Specifications
- Catalog No:
- 20413-100UL
- 20413-500UL
- Antigen symbol:
- IgG
- IgG
- Antigen name:
- Rat IgG
- Rat IgG
- Antibody type:
- Secondary
- Secondary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- Reactivity:
- Rat
- Rat
- Host:
- Goat
- Goat
- Form:
- Liquid
- Liquid
- Purification:
- Affinity purified
- Affinity purified
- Concentration:
- 0,5 mg/ml
- 0,5 mg/ml
- Cross adsorption:
- Bovine, horse, human, rabbit
- Bovine, horse, human, rabbit
- Storage buffer:
- PBS, 50% glycerol, 2 mg/ml BSA, 0,05% sodium azide
- PBS, 50% glycerol, 2 mg/ml BSA, 0,05% sodium azide
- Storage temperature:
- Store at –20 °C.
- Store at –20 °C.
Specifications
Frequently Bought Together