Order Entry
Netherlands
ContactUsLinkComponent
Anti-IgG Goat Polyclonal Antibody (APC (Allophycocyanin))
Anti-IgG Goat Polyclonal Antibody (APC (Allophycocyanin))
Supplier:  Biotium
Certificates
Anti-IgG Goat Polyclonal Antibody (APC (Allophycocyanin))
Supplier:  Biotium

Specifications

  • Catalog No:
  • 20411-500UL
  • 20411-100UL
  • Antigen symbol:
  • IgG
  • IgG
  • Antigen name:
  • Mouse IgG
  • Mouse IgG
  • Antibody type:
  • Secondary
  • Secondary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Goat
  • Goat
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Affinity purified
  • Affinity purified
  • Concentration:
  • 0,5 mg/ml
  • 0,5 mg/ml
  • Cross adsorption:
  • No
  • No
  • Storage buffer:
  • PBS, 50% glycerol, 2 mg/ml BSA, 0,05% sodium azide
  • PBS, 50% glycerol, 2 mg/ml BSA, 0,05% sodium azide
  • Storage temperature:
  • Store at –20 °C.
  • Store at –20 °C.

Specifications

Frequently Bought Together