Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-IgA Goat Polyclonal Antibody (CF™ 594)
 :  Biotium
 
Anti-IgA Goat Polyclonal Antibody (CF™ 594)
 :  Biotium

 

  • Catalog No:
  • BTIU20429-500UL
  • BTIU20429-50UL
  • Antigen symbol:
  • IgA
  • IgA
  • Antigen name:
  • Human IgA
  • Human IgA
  • Antibody type:
  • Secondary
  • Secondary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • CF® 594
  • CF® 594
  • Reactivity:
  • Human
  • Human
  • Host:
  • Goat
  • Goat
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Affinity purified
  • Affinity purified
  • Concentration:
  • 2 mg/mL
  • 2 mg/mL
  • Cross adsorption:
  • Human IgG
  • Human IgG
  • Storage buffer:
  • PBS, 50% glycerol, 2 mg/ml BSA, 0,05% sodium azide
  • PBS, 50% glycerol, 2 mg/ml BSA, 0,05% sodium azide
  • Storage temperature:
  • Store at –20 °C.
  • Store at –20 °C.

 

Frequently Bought Together