Specifications
- Antibody type:Secondary
- Antigen name:Human IgG, Fc Gamma
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:CF® 640R
- Host:Goat
- Reactivity:Human
- Cross adsorption:Bovine, horse, mouse, rabbit
- Form:Lyophilized
- Storage buffer:PBS, 15 mg/ml BSA, 20 mg/mL trehalose (after reconstitution)
- Storage temperature:Store at –20 °C.
- Concentration:2 mg/ml (after reconstitution)
- Purification:Affinity purified
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Goat anti-human IgG, Fc gamma specific antibody reacts specifically with the human IgG heavy (γ) chains and not with light chains and F(ab’)2 fragment on all human immunoglobulins. To minimize cross-reactivity, it has been adsorbed against bovine, horse, mouse and rabbit serum proteins prior to conjugation. CF™640R (Ex/Em 642/662 nm) is a novel far-red rhodamine-based dye spectrally similar to Cy®5 and Alexa Fluor® 647. CF™640R is much brighter than Cy®5 and at least as bright as Alexa Fluor® 647 when excited at 633 or 635 nm. However, the most important advantage of CF™640R is its exceptional photostability.
Alexa Fluor® is a registered trademark of Thermo Fisher Scientific. Cy®5 is a registered trademark of GE Healthcare.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF640R
Epitope:
Host: Goat
Isotype:
Reactivity: Human