Anti-EAAT1 Antibody
Catalog # BSBTPB9959
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 1 (glial high affinity glutamate transporter), member 3
- Antigen symbol:Solute carrier family 1, member 3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:6507
- Antigen synonyms:EA6|GLAST|GLAST1|EAAT1
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Excitatory amino acid transporter 1(SLC1A3) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Type: Primary
Antigen: SLC1A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat