Rat IgG2A Isotype Control
Catalog # SANTSC-3883
Supplier: Santa Cruz Biotechnology
Specifications
- Type antilichaam:Controle
- antigeen symbool:IgG2a
- Conjugatie:Onconjugeerd
- Flowcytometrie:Yes
- Isotype:IgG2a
- VPE:100 Tests
Specifications
About this item
Type: Control
Antigen:
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: