L-Tryptophan ethyl ester hydrochloride, powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:2899-28-7
:
Formula:C₁₃H₁₆N₂O₂
MDL Number:MFCD00038993
:268.74 g/mol
Specification Test Results
- Form:Light Gray Powder
- Storage:0 °C, desiccate
- Optical Rotation:[α]20/D +10 ±1°, c = 2% in H2O
Specification Test Results
Frequently Bought Together