


L-α-Glycerophosphoryl Choline, Cadmium Chloride Complex (from egg yolk) ∼98%, white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:64681-08-9
:
Formula:C₈H₂₀CdCl₂NO₆P
:440.537 g/mol
EINECS:265-010-1
:GHS06
:GHS08
:GHS09
Storage Temperature:Freezer
Frequently Bought Together