Combination pH electrodes, sensION™+
Supplier: Hach
Certificates
About this item
3-in-1 laboratory combination pH electrodes with built-in temperature sensor.
- 5010T and 5011T are low maintenance electrodes for general applications
- 5014T and 5021T are high performance electrodes for special or difficult applications
- For use with Hach sensION+ laboratory pH meters
Delivery: Supplied with basic user manual. 5014T and 5021T also include 3,0 M KCl electrolyte refill solution (30 ml).
Frequently Bought Together