Anti-DUSP1 Mouse Monoclonal Antibody [clone: D-3]
Catalog # SANTSC-271684
Supplier: Santa Cruz Biotechnology
Specifications
- Antibody type:Primary
- Antigen name:Dual specificity phosphatase 1
- Antigen symbol:DUSP1
- Clonality:Monoclonal
- Clone:D-3
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG3
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Gene ID:1843
- Antigen synonyms:CL100|PTPN10|HVH1|MKP-1|MKP1
- Amino acid number:333 to 366
- Molecular weight:40 kDa
- Storage temperature:+4 °C
- Size:200 µg/ml
- Pk:200 µG
Specifications
About this item
Type: Primary
Antigen: DUSP1 (dual specificity phosphatase 1)
Clonality: Monoclonal
Clone: D-3
Conjugation:
Epitope: C-terminal
Host: Mouse
Isotype: IgG3
Reactivity: Human, Mouse, Rat