Anti-MYH7 Mouse Monoclonal Antibody [clone: A4.951]
Catalog # SANTSC-53090
Supplier: Santa Cruz Biotechnology
Specifications
- Antibody type:Primary
- Antigen name:myosin, heavy chain 7, cardiac muscle, beta
- Antigen symbol:MYH7
- Clonality:Monoclonal
- Clone:A4.951
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Fish,Rabbit,Human,Mouse
- Western blot:Yes
- Gene ID:4625
- Antigen synonyms:SPMD|CMH1|SPMM|MYHCB|MPD1|CMD1S
- Molecular weight:223 kDa
- Storage temperature:+4 °C
- Size:200 µg/ml
- Pk:1
Specifications
About this item
Type: Primary
Antigen: MYH7 (myosin, heavy chain 7, cardiac muscle, beta)
Clonality: Monoclonal
Clone: A4.951
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Fish, Human, Mouse, Rabbit