Sorbic acid, powder NF
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:110-44-1
:
Formula:CH₃CH=CHCH=CHCOOH
MDL Number:MFCD00002703
:112.13 g/mol
Merck Index:13,08793
Boiling Pt:228 °C (1013 hPa)
Melting Pt:132…135 °C
Density:1.204 g/cm³ (20 °C)
Flash Pt:127 °C (closed cup)
EINECS:203-768-7
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay:99% Min.
Specification Test Results
Frequently Bought Together