Order Entry
Netherlands
ContactUsLinkComponent
4-Chloro-2-(trifluoromethyl)phenyl isocyanate
4-Chloro-2-(trifluoromethyl)phenyl isocyanate
Catalog # L09850.03
CAS Number:  
4-Chloro-2-(trifluoromethyl)phenyl isocyanate
Catalog # L09850.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :221.57 g/mol
Boiling Pt:85 - 90 °C (13 -rr)
Density:1.4666 g/cm³ (20 °C)
ADR:6.1,III
CAS Number:16588-69-5
Formula:C₈H₃ClF₃NO
MDL Number:MFCD00013873