
Dimethylaminoacetyl chloride hydrochloride ≥85%, tech.
Catalog # L18475.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
About this item
CAS Number:60853-81-8
:
Formula:(CH₃)₂NCH₂COCl·HCl
MDL Number:MFCD02094025
:158.03 g/mol
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS05