Order Entry
Netherlands
ContactUsLinkComponent
4,4,4-Trifluorocrotononitrile 96%
Toxic
Irritant
4,4,4-Trifluorocrotononitrile 96%
Catalog # L16870.03
CAS Number:  
Toxic
Irritant
4,4,4-Trifluorocrotononitrile 96%
Catalog # L16870.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

CAS Number:406-86-0
 :
Formula:CF₃CH=CHCN
MDL Number:MFCD00190645
 :121.06 g/mol
Boiling Pt:84…85 °C
ADR:6.1,II
 :GHS06
 :GHS07
Storage Temperature:Ambient