
Perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecanoyl fluoride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:27639-98-1
:
Formula:C₁₂F₂₄O₄
MDL Number:MFCD00084743
:664.09 g/mol
Boiling Pt:158…161 °C
Density:1.80
Storage Temperature:Ambient
ADR:8,II
:GHS05
Related Information
Frequently Bought Together