Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Boranepyridine complex 95%

Boranepyridine complex 95%

50 g
Catalogus # L13178.18
Toxic
Irritant
Leverancier:   Thermo Fisher Scientific
Boranepyridine complex 95%
Catalogus # L13178.18
CAS-nummer:  110-51-0
Boranepyridine complex 95%
Boranepyridine complex 95%

50 g

CAS-nummer:  110-51-0
Price
166,00 €
Log in for availability
1 * 50 g
Product Details & Documents

Purity: 95% CAS No.: 110-51-0 Molecular Formula: C5H8BN Formula Weight: 92.93 Storage Temperature: 2°C to 8°C Flash Point: 73°(163°F) Refractive Index: 1.5320 Odour: Unpleasant Melting Point: 9-11° Boiling Point: 65°/1mm Beilstein Reference: 3674913 UN No.: UN2810 MDL No.: MFCD00012435

Synonyms:  Pyridine Borane, Borane, pyridine complex
  • CAS Number:110-51-0
  • Formula:C₅H₈BN
  • MDL Number:MFCD00012435
  • Molecular Weight:92.94 g/mol
  • Boiling Pt:65 °C (1 mmHg)
  • Melting Pt:9…11 °C
  • Density:0.920
  • Flash Pt:73 °C (163 °F)
  • EINECS:203-773-4
  • ADR:6.1,II
Specifications

Specifications

  • Pk:
    50 g

Specifications