1-(2-Naphthyl)ethanol 97%
Catalog # L05108.09
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:10 g
Specifications
About this item
:
:172.23 g/mol
Boiling Pt:170 °C (13 mmHg)
Melting Pt:72…75 °C
Storage Temperature:Ambient
EINECS:230-630-3
CAS Number:7228-47-9
Formula:C₁₂H₁₂O
MDL Number:MFCD00004111