
Flucloxacillin sodium
Catalog # J66615.MC
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:100 mg
Specifications
About this item
:
:476.859 g/mol
Boiling Pt:677 °C
Melting Pt:176…178 °C
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:217-428-0
:GHS08
CAS Number:1847-24-1
Formula:C₁₉H₁₇ClFN₃NaO₅S
MDL Number:MFCD01682108