Order Entry
Netherlands
ContactUsLinkComponent
5'-O-(4,4'-Dimethoxytrityl)-N-2-isobutyryl-2'-O-methylguanosine-3'-(2-cyanoethyl diisopropylphosphoramidite) 98%
Irritant
5'-O-(4,4'-Dimethoxytrityl)-N-2-isobutyryl-2'-O-methylguanosine-3'-(2-cyanoethyl diisopropylphosphoramidite) 98%
Catalog # J65100.03
CAS Number:  
Irritant
5'-O-(4,4'-Dimethoxytrityl)-N-2-isobutyryl-2'-O-methylguanosine-3'-(2-cyanoethyl diisopropylphosphoramidite) 98%
Catalog # J65100.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :869.94 g/mol
 :GHS07
Storage Temperature:Freezer
Formula:C₄₅H₅₆N₇O₉P
MDL Number:MFCD00792625