2'-Deoxyadenosine-5'-monophosphate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:653-63-4
:
Formula:C₁₀H₁₄N₅O₆P
MDL Number:MFCD00005753
:331.22 g/mol
EINECS:211-503-1
Related Information
Frequently Bought Together