Imidazole-buffered saline (5X)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS No.: 288-32-4
Physical Form: Liquid
Odour: Odourless
MDL No.: MFCD00005183
Specifications
- CAS No.:288-32-4
- MDL No.:MFCD00005183
- Physical form:Liquid
- Odour/scent:Odourless
Specifications
Related Information
Frequently Bought Together