MES-buffered saline (5X), pH 6.5
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS No.: 145224-94-8
Physical Form: Liquid
MDL No.: MFCD00283848
Specifications
- CAS No.:145224-94-8
- MDL No.:MFCD00283848
- Physical form:Liquid
Specifications
Related Information
Frequently Bought Together