Order Entry
Netherlands
ContactUsLinkComponent
XTT sodium salt
XTT sodium salt
Catalog # J61726.MD
CAS Number:  
XTT sodium salt
Catalog # J61726.MD
CAS Number:  

Specifications

  • Pk:
    250 mg

Specifications

About this item

 :
 :673.52 g/mol
CAS Number:111072-31-2
Formula:C₂₂H₁₇N₇O₁₃S₂Na
MDL Number:MFCD00083517