H-7 dihydrochloride
Catalog # J61697.MA
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- VPE:10 mg
- Seq:2275
Specifications
About this item
:
:364.29 g/mol
Melting Pt:191…193 °C
Storage Temperature:Koelkast
CAS Number:108930-17-2
Formula:C₁₄H₁₇N₃O₂S·2HCl
MDL Number:MFCD00036961