Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dde-Lys(Fmoc)-OH
Dde-Lys(Fmoc)-OH 98%
Dde-Lys(Fmoc)-OH 98%
Dde-Lys(Fmoc)-OH 98%
Catalog # H66446.03
Supplier:
Thermo Fisher Scientific
CAS Number:
Certificates
Expand image
Dde-Lys(Fmoc)-OH 98%
Catalog # H66446.03
Supplier:
Thermo Fisher Scientific
CAS Number:
Certificates
Specifications
Pk:
1 g
Specifications
About this item
Product Details
Documentation
:
:
532.637 g/mol
CAS Number:
156648-40-7
Formula:
C₃₁H₃₆N₂O₆
MDL Number:
MFCD00467662
Product Details
Documentation