
5-Hydroxy-L-tryptophan hydrate 98%
Catalog # A13954.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:4350-09-8
:
Formula:C₁₁H₁₂N₂O₃
MDL Number:MFCD00064341
:220.23 g/mol (anhy)
Merck Index:14,4847
Melting Pt:258 °C (dec.)
Storage Temperature:Refrigerator
EINECS:224-411-1
UN:2811
ADR:6.1,III
:GHS07
Formula:C₁₁H₁₂N₂O₃·xH₂O