
Methyl (S)-(+)-2-pyrrolidone-5-carboxylate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4931-66-2
:
Formula:C₆H₉NO₃
MDL Number:MFCD00080931
:143.14 g/mol
Boiling Pt:90 °C (0.3 mmHg)
Density:1.226
Flash Pt:113 °C (235 °F)
EINECS:225-567-3
:GHS07
Storage Temperature:Ambient
Frequently Bought Together