Order Entry
Netherlands
ContactUsLinkComponent
Trimethylboroxine 50% (w/w) in THF ≥98%
Flammable
Corrosive
Health
Irritant
Trimethylboroxine 50% (w/w) in THF ≥98%
Catalog # H61464.AA
CAS Number:  
Flammable
Corrosive
Health
Irritant
Trimethylboroxine 50% (w/w) in THF ≥98%
Catalog # H61464.AA
CAS Number:  

Specifications

  • Pk:
    5 mL

Specifications

About this item

CAS Number:823-96-1
 :
Formula:C₃H₉B₃O₃
MDL Number:MFCD00013354
 :125.532 g/mol
Density:0.894
Storage Temperature:Ambient
UN:2924
ADR:3,II
 :GHS02
 :GHS05
 :GHS08
 :GHS07