
O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium hexafluorophosphate ≥98%
Catalog # H54035.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:359.21 g/mol
Melting Pt:∼ 220 °C (dec.)
:GHS07
CAS Number:265651-18-1
Formula:C₉H₁₆F₆N₃O₃P
MDL Number:MFCD01863753