Order Entry
Netherlands
ContactUsLinkComponent
 
Irritant
2-(4-Pyridyl)-7-azabenzimidazole 97%
Catalog # H52213.MD
CAS Number:  
Irritant
2-(4-Pyridyl)-7-azabenzimidazole 97%
Catalog # H52213.MD
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

CAS Number:63411-78-9
 :
Formula:C₁₁H₈N₄
MDL Number:MFCD01001152
 :196.21 g/mol
Melting Pt:240…248 °C
 :GHS07
Storage Temperature:Ambient