

n-Propylmagnesium chloride 1 M in MeTHF (2-methyltetrahydrofuran)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2234-82-4
:
Formula:C₃H₇ClMg
MDL Number:MFCD00000474
:102.844 g/mol
UN:3399
ADR:4.3,II
:GHS02
:GHS05
Storage Temperature:Ambient
Related Information
Frequently Bought Together