Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(3-Bromophenyl)-1H-pyrazole
3-(3-Bromophenyl)-1H-pyrazole 98%
3-(3-Bromophenyl)-1H-pyrazole 98%
H50438.06
:
Thermo Fisher Scientific
:
3-(3-Bromophenyl)-1H-pyrazole 98%
H50438.06
:
Thermo Fisher Scientific
:
Pk:
5 g
Seq:
3173
:
:
223.07 g/mol
:
Ambient
:
149739-65-1
:
C₉H₇BrN₂
:
MFCD01940433