
Methyl-1H-pyrazole-3-carboxylate 97%
Catalog # H34148.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
Specifications
About this item
:
Melting Pt:141…143 °C
:126.11 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:15366-34-4
Formula:C₅H₆N₂O₂
MDL Number:MFCD00649381