
4-Chloro-5-(4-chlorophenyl)thieno[2,3-d]pyrimidine 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:331761-46-7
:
Formula:C₁₂H₆Cl₂N₂S
MDL Number:MFCD01038375
:281.16 g/mol
Melting Pt:139.5…141 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together