
1-(2-Tetrahydropyranyl)-1H-imidazole-5-boronic acid pinacol ester 95%
Catalog # H32369.ME
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:278.16 g/mol
Melting Pt:129…133 °C
:GHS07
CAS Number:1029684-37-4
Formula:C₁₄H₂₃BN₂O₃
MDL Number:MFCD11100960