
3-(4-Nitrophenyl)-1H-pyrazole 97%
Catalog # H32332.04
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:2 g
- Seq:3107
Specifications
About this item
CAS Number:20583-31-7
:
Formula:C₉H₇N₃O₂
MDL Number:MFCD00665859
:189.18 g/mol
Melting Pt:194…195 °C
:GHS07
Storage Temperature:Ambient